Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttc25
Entrez Gene ID
303534
Gene Name
tetratricopeptide repeat domain 25
Gene Aliases
RGD1311859
Chromosome Location
Chr.10: 88458636 - 88488237 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107049.1 NP_001100519.1 4 - 5 587 67
XM_006247310.2 XP_006247372.1 4 - 5 1441 67
GenBank mRNA BC162060.1 - 4 - 5 530 67

View Full Product Details