Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhbdl1
Entrez Gene ID
117025
Gene Name
rhomboid, veinlet-like 1 (Drosophila)
Gene Aliases
Rhbdl
Chromosome Location
Chr.10: 15201503 - 15205885 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191822.1 NP_001178751.1 5 - 6 744 124
XM_006245894.2 XP_006245956.1 4 - 5 2324 124
XM_008767537.1 XP_008765759.1 5 - 6 2498 124
GenBank mRNA Y17258.1 - 2 - 3 128 124

View Full Product Details