Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsad1
Entrez Gene ID
688146
Gene Name
radical S-adenosyl methionine domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 82269104 - 82276599 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271211.1 NP_001258140.1 - 1495 125

View Full Product Details