Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dok2
Entrez Gene ID
290361
Gene Name
docking protein 2
Gene Aliases
Dok-2
Chromosome Location
Chr.15: 52450150 - 52455414 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106048.1 NP_001099518.1 3 - 4 553 61
XM_006252262.1 XP_006252324.1 4 - 5 1322 61
XM_006252264.2 XP_006252326.1 4 - 5 1783 61
XM_008770826.1 XP_008769048.1 3 - 4 2757 61

View Full Product Details