Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc15
Entrez Gene ID
290370
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 15
Gene Aliases
Dnajd1
Chromosome Location
Chr.15: 59990131 - 60056710 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106050.1 NP_001099520.1 4 - 5 469 109
GenBank mRNA FQ229062.1 - 4 - 5 343 109

View Full Product Details