Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpc5
Entrez Gene ID
306157
Gene Name
glypican 5
Gene Aliases
-
Chromosome Location
Chr.15: 100283131 - 101776838 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107285.2 NP_001100755.2 1 - 2 166 148

View Full Product Details