Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cldn10
Entrez Gene ID
290485
Gene Name
claudin 10
Gene Aliases
-
Chromosome Location
Chr.15: 104026589 - 104115748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106058.1 NP_001099528.1 5 - 6 981 87
XM_006252450.2 XP_006252512.1 4 - 5 822 87
XM_006252451.2 XP_006252513.1 4 - 5 714 87

View Full Product Details