The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022868.1 | NP_074059.1 | 2 - 3 | 2544 | 127 |
XM_006252375.2 | XP_006252437.1 | 2 - 3 | 3501 | 127 | |
XM_008770897.1 | XP_008769119.1 | 3 - 4 | 3264 | 127 | |
GenBank mRNA | AB026154.1 | - | 2 - 3 | 2544 | 127 |