The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017254.1 | NP_058950.1 | 2 - 3 | 678 | 77 |
GenBank mRNA | M30705.1 | - | 2 - 3 | 678 | 77 |
M64867.1 | - | - | 1084 | 77 | |
X13971.1 | - | - | 1221 | 77 |