Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Entpd4
Entrez Gene ID
361063
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Gene Aliases
-
Chromosome Location
Chr.15: 51222954 - 51250787 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108384.1 NP_001101854.1 4 - 5 623 59

View Full Product Details