Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Entpd4
Entrez Gene ID
361063
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Gene Aliases
-
Chromosome Location
Chr.15: 51222954 - 51250787 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108384.1 NP_001101854.1 6 - 7 961 86
XM_006252275.2 XP_006252337.1 11 - 12 1714 86
XM_006252276.2 XP_006252338.1 11 - 12 2117 86
XM_006252277.2 XP_006252339.1 11 - 12 1690 86

View Full Product Details