Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cib4
Entrez Gene ID
688819
Gene Name
calcium and integrin binding family member 4
Gene Aliases
-
Chromosome Location
Chr.6: 27241804 - 27303617 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001068424.4 XP_001068424.2 1 - 2 240 66

View Full Product Details