Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cib4
Entrez Gene ID
688819
Gene Name
calcium and integrin binding family member 4
Gene Aliases
-
Chromosome Location
Chr.6: 27241804 - 27303617 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001068424.4 XP_001068424.2 4 - 5 510 116

View Full Product Details