Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sav1
Entrez Gene ID
299116
Gene Name
salvador family WW domain containing protein 1
Gene Aliases
-
Chromosome Location
Chr.6: 92376470 - 92398678 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097581.1 NP_001091050.1 3 - 4 982 112
GenBank mRNA AM261215.1 - 3 - 4 982 112
BC161803.1 - 3 - 4 829 112

View Full Product Details