Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Matn3
Entrez Gene ID
313954
Gene Name
matrilin 3
Gene Aliases
-
Chromosome Location
Chr.6: 34071428 - 34091048 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108013.1 NP_001101483.1 6 - 7 1393 97

View Full Product Details