Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc7a15
Entrez Gene ID
298873
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 15
Gene Aliases
RGD1560726
Chromosome Location
Chr.6: 33728107 - 33739115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106714.1 NP_001100184.1 5 - 6 1252 104

View Full Product Details