Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tssc1
Entrez Gene ID
362721
Gene Name
tumor suppressing subtransferable candidate 1
Gene Aliases
-
Chromosome Location
Chr.6: 48021771 - 48135241 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012192.1 NP_001012192.1 3 - 4 333 61
GenBank mRNA BC087633.1 - 3 - 4 333 61

View Full Product Details