The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106717.1 | NP_001100187.1 | 3 - 4 | 592 | 67 |
XM_006239896.1 | XP_006239958.1 | 2 - 3 | 419 | 67 |