The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013047.1 | NP_001013065.1 | 5 - 6 | 379 | 80 |
XM_008764483.1 | XP_008762705.1 | 4 - 5 | 412 | 80 | |
XM_008764484.1 | XP_008762706.1 | 4 - 5 | 412 | 80 | |
GenBank mRNA | BC079400.1 | - | 5 - 6 | 526 | 80 |
BC091225.1 | - | 5 - 6 | 379 | 80 | |
FQ224358.1 | - | 4 - 5 | 417 | 80 |