Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adcy3
Entrez Gene ID
64508
Gene Name
adenylate cyclase 3
Gene Aliases
-
Chromosome Location
Chr.6: 28571133 - 28648849 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130779.2 NP_570135.2 14 - 15 2859 102
XM_006239847.2 XP_006239909.1 13 - 14 2595 102
XM_006239848.2 XP_006239910.1 13 - 14 2598 102
XM_006239849.2 XP_006239911.1 13 - 14 2592 102
XM_006239850.2 XP_006239912.1 13 - 14 2601 102
XM_008764532.1 XP_008762754.1 14 - 15 2715 102
GenBank mRNA M55075.1 - 14 - 15 2784 102

View Full Product Details