Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adcy3
Entrez Gene ID
64508
Gene Name
adenylate cyclase 3
Gene Aliases
-
Chromosome Location
Chr.6: 28571133 - 28648849 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130779.2 NP_570135.2 17 - 18 3173 84
XM_006239847.2 XP_006239909.1 16 - 17 2909 84
XM_006239848.2 XP_006239910.1 16 - 17 2912 84
XM_006239849.2 XP_006239911.1 16 - 17 2906 84
XM_006239850.2 XP_006239912.1 16 - 17 2912 84
XM_008764532.1 XP_008762754.1 17 - 18 3029 84
GenBank mRNA M55075.1 - 17 - 18 3098 84

View Full Product Details