Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Haao
Entrez Gene ID
56823
Gene Name
3-hydroxyanthranilate 3,4-dioxygenase
Gene Aliases
-
Chromosome Location
Chr.6: 7045145 - 7059219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020076.2 NP_064461.1 9 - 10 870 99
XM_006239684.2 XP_006239746.1 8 - 9 1706 99
GenBank mRNA BC085739.1 - 9 - 10 824 99
D44494.1 - 9 - 10 871 99
FQ219702.1 - 9 - 10 884 99

View Full Product Details