Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc2
Entrez Gene ID
299113
Gene Name
kelch domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 91595647 - 91610980 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034133.1 NP_001029305.1 7 - 8 889 101
GenBank mRNA BC105756.1 - 7 - 8 889 101

View Full Product Details