The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034133.1 | NP_001029305.1 | 8 - 9 | 950 | 101 |
GenBank mRNA | BC105756.1 | - | 8 - 9 | 950 | 101 |