Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl28
Entrez Gene ID
299103
Gene Name
kelch-like family member 28
Gene Aliases
Btbd5
Chromosome Location
Chr.6: 86680826 - 86713370 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106735.1 NP_001100205.1 1 - 2 128 103

View Full Product Details