Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim54
Entrez Gene ID
362708
Gene Name
tripartite motif-containing 54
Gene Aliases
Rnf30
Chromosome Location
Chr.6: 26603364 - 26623950 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013217.1 NP_001013235.1 1 - 2 358 55
GenBank mRNA BC083706.1 - 1 - 2 358 55

View Full Product Details