Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nova1
Entrez Gene ID
298992
Gene Name
neuro-oncological ventral antigen 1
Gene Aliases
Nova-1
Chromosome Location
Chr.6: 66960126 - 67085844 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100541.1 NP_001094011.1 1 - 2 137 127
XM_006240069.2 XP_006240131.1 1 - 2 591 127
GenBank mRNA AY262017.1 - 1 - 2 137 127

View Full Product Details