Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx5
Entrez Gene ID
287765
Gene Name
DEAD (Asp-Glu-Ala-Asp) box helicase 5
Gene Aliases
-
Chromosome Location
Chr.10: 94979759 - 94988461 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007613.1 NP_001007614.1 13 - 14 1670 121
XM_008768312.1 XP_008766534.1 12 - 13 1659 121
XM_008768313.1 XP_008766535.1 12 - 13 1582 121
GenBank mRNA BC079036.1 - 13 - 14 1670 121
FJ168767.1 - 12 - 13 1501 121
FQ227563.1 - 1 - 2 205 121

View Full Product Details