Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Socs3
Entrez Gene ID
89829
Gene Name
suppressor of cytokine signaling 3
Gene Aliases
Cish3, Socs-3, Ssi-3
Chromosome Location
Chr.10: 106973863 - 106976969 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768398.1 XP_008766620.1 1 - 2 295 110

View Full Product Details