Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myh2
Entrez Gene ID
691644
Gene Name
myosin, heavy chain 2, skeletal muscle, adult
Gene Aliases
MyHC-2A, MyHC-IIA, Myh1, Myh2a
Chromosome Location
Chr.10: 53711895 - 53738164 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135157.1 NP_001128629.1 - 2781 120

View Full Product Details