Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Copz2
Entrez Gene ID
360611
Gene Name
coatomer protein complex, subunit zeta 2
Gene Aliases
-
Chromosome Location
Chr.10: 84718824 - 84731880 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108294.1 NP_001101764.1 2 - 3 187 104
GenBank mRNA FQ225734.1 - 2 - 3 191 104

View Full Product Details