The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107072.1 | NP_001100542.1 | 4 - 5 | 1447 | 59 |
XM_008768476.1 | XP_008766698.1 | 4 - 5 | 6673 | 59 |