Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnip1
Entrez Gene ID
65023
Gene Name
Kv channel-interacting protein 1
Gene Aliases
Kchip1
Chromosome Location
Chr.10: 18555313 - 18942677 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261387.1 NP_001248316.1 2 - 3 257 69
NM_001261389.1 NP_001248318.1 2 - 3 93 69
GenBank mRNA AY082657.1 - 2 - 3 93 69
AY142709.1 - 2 - 3 93 69

View Full Product Details