Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxk2
Entrez Gene ID
303753
Gene Name
forkhead box K2
Gene Aliases
-
Chromosome Location
Chr.10: 110468893 - 110518854 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107075.1 NP_001100545.1 3 - 4 469 106
XM_006247890.2 XP_006247952.1 3 - 4 866 106
XM_006247891.2 XP_006247953.1 3 - 4 866 106
GenBank mRNA BC168919.1 - 2 - 3 334 106

View Full Product Details