The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012357.1 | NP_001012357.1 | 2 - 3 | 243 | 63 |
GenBank mRNA | AY863011.1 | - | 2 - 3 | 243 | 63 |
BC089865.1 | - | 2 - 3 | 243 | 63 | |
EF121990.1 | - | 2 - 3 | 135 | 63 | |
EF121991.1 | - | 2 - 3 | 135 | 63 |