Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt24
Entrez Gene ID
287675
Gene Name
keratin 24, type I
Gene Aliases
Ka24
Chromosome Location
Chr.10: 87190184 - 87195075 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004131.1 NP_001004131.1 4 - 5 943 77

View Full Product Details