Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tubg1
Entrez Gene ID
252921
Gene Name
tubulin, gamma 1
Gene Aliases
-
Chromosome Location
Chr.10: 89030865 - 89037455 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145778.2 NP_665721.1 1 - 2 159 96
GenBank mRNA BC070957.1 - 1 - 2 159 96

View Full Product Details