Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmd
Entrez Gene ID
303439
Gene Name
monocyte to macrophage differentiation-associated
Gene Aliases
Maf
Chromosome Location
Chr.10: 77755178 - 77783739 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007673.1 NP_001007674.1 2 - 3 300 60
XM_006247126.1 XP_006247188.1 2 - 3 338 60
XM_006247127.2 XP_006247189.1 2 - 3 1160 60
GenBank mRNA BC081965.1 - 2 - 3 300 60

View Full Product Details