Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Shpk
Entrez Gene ID
287479
Gene Name
sedoheptulokinase
Gene Aliases
Carkl
Chromosome Location
Chr.10: 59765328 - 59789676 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033682.1 NP_001028854.1 3 - 4 524 61
GenBank mRNA BC101914.1 - 3 - 4 524 61

View Full Product Details