Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clk4
Entrez Gene ID
287269
Gene Name
CDC-like kinase 4
Gene Aliases
-
Chromosome Location
Chr.10: 36715565 - 36733476 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013041.1 NP_001013059.1 10 - 11 1153 67
XM_006246234.2 XP_006246296.1 11 - 12 1363 67
XM_006246236.1 XP_006246298.1 14 - 15 1566 67
XM_006246237.1 XP_006246299.1 12 - 13 1268 67
XM_006246238.2 XP_006246300.1 11 - 12 2783 67
XM_008767670.1 XP_008765892.1 12 - 13 2237 67
XM_008767671.1 XP_008765893.1 11 - 12 2159 67
XM_008767672.1 XP_008765894.1 11 - 12 1202 67
XM_008767673.1 XP_008765895.1 10 - 11 2855 67
XM_008767674.1 XP_008765896.1 9 - 10 2642 67
XM_008767675.1 XP_008765897.1 12 - 13 1429 67
XM_008767676.1 XP_008765898.1 10 - 11 1154 67
GenBank mRNA BC079006.1 - 10 - 11 1153 67
FQ223275.1 - 13 - 14 3213 67

View Full Product Details