Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf43
Entrez Gene ID
303412
Gene Name
ring finger protein 43
Gene Aliases
RGD1305204
Chromosome Location
Chr.10: 74956889 - 75028951 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135921.1 NP_001129393.1 3 - 4 451 72
XM_006247090.2 XP_006247152.1 4 - 5 1936 72
XM_006247091.2 XP_006247153.1 4 - 5 1711 72
XM_006247092.2 XP_006247154.1 3 - 4 2456 72
XM_006247094.2 XP_006247156.1 2 - 3 1812 72
XM_008768038.1 XP_008766260.1 3 - 4 2847 72
XM_008768040.1 XP_008766262.1 2 - 3 2392 72
XM_008768041.1 XP_008766263.1 2 - 3 2040 72

View Full Product Details