Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnmd
Entrez Gene ID
64104
Gene Name
tenomodulin
Gene Aliases
ChM1L
Chromosome Location
Chr.X: 104684676 - 104700173 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022290.1 NP_071626.1 1 - 2 269 114
GenBank mRNA AB055423.1 - 1 - 2 58 114
AF191769.1 - 1 - 2 269 114

View Full Product Details