Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Satl1
Entrez Gene ID
302325
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Gene Aliases
RGD1565932
Chromosome Location
Chr.X: 83772526 - 83788430 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163543.1 NP_001157015.1 4 - 5 2062 75

View Full Product Details