Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bspry
Entrez Gene ID
64027
Gene Name
B-box and SPRY domain containing
Gene Aliases
-
Chromosome Location
Chr.5: 78334284 - 78356462 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022261.2 NP_071597.2 1 - 2 375 103
GenBank mRNA BC062051.1 - 1 - 2 323 103
BC166587.1 - 1 - 2 375 103

View Full Product Details