The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108024.1 | NP_001101494.1 | 5 - 6 | 2623 | 82 |
XM_008764687.1 | XP_008762909.1 | 3 - 4 | 2223 | 82 |