Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nampt
Entrez Gene ID
297508
Gene Name
nicotinamide phosphoribosyltransferase
Gene Aliases
Pbef, Pbef1
Chromosome Location
Chr.6: 52122085 - 52156473 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177928.3 NP_808789.1 1 - 2 86 120
GenBank mRNA AB081730.1 - 1 - 2 61 120
AY831728.1 - 1 - 2 59 120
BC085681.1 - 1 - 2 86 120

View Full Product Details