Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mpp5
Entrez Gene ID
314259
Gene Name
membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5)
Gene Aliases
-
Chromosome Location
Chr.6: 101923670 - 102029705 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108034.1 NP_001101504.1 12 - 13 1992 138
XM_006240258.2 XP_006240320.1 13 - 14 2561 138
XM_008764773.1 XP_008762995.1 13 - 14 2165 138
XM_008764774.1 XP_008762996.1 13 - 14 1929 138
GenBank mRNA BC168247.1 - 13 - 14 2050 138

View Full Product Details