The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008320.1 | NP_001008321.1 | 3 - 4 | 661 | 74 |
GenBank mRNA | BC086525.1 | - | 3 - 4 | 661 | 74 |