The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270619.1 | NP_001257548.1 | 3 - 4 | 1368 | 115 |
NM_001270620.1 | NP_001257549.1 | 4 - 5 | 1842 | 115 | |
NM_033096.2 | NP_149087.1 | 3 - 4 | 1368 | 115 | |
XM_008764454.1 | XP_008762676.1 | 2 - 3 | 979 | 115 | |
GenBank mRNA | AJ271834.1 | - | 3 - 4 | 1361 | 115 |
AJ271837.1 | - | 3 - 4 | 1361 | 115 | |
BC061986.1 | - | 3 - 4 | 1323 | 115 | |
BC081762.1 | - | 4 - 5 | 1842 | 115 | |
FM075941.1 | - | 1 - 2 | 108 | 115 | |
FM108050.1 | - | 2 - 3 | 523 | 115 | |
S90449.1 | - | 3 - 4 | 1654 | 115 |