Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vti1b
Entrez Gene ID
100359512
Gene Name
vesicle transport through interaction with t-SNAREs 1B
Gene Aliases
-
Chromosome Location
Chr.6: 102340260 - 102353598 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750196.3 XP_003750244.1 3 - 4 682 80

View Full Product Details